CAS RN 274694-98-3
CAS RN 274694-98-3
IUPAC Name:
11-methyl-11,21-diazatetracyclo[12.7.0.0³,⁸.0¹⁵,²⁰]henicosa-1(14),3,5,7,15,17,19-heptaene
Synonyms:
11-methyl-11,21-diazatetracyclo[12.7.0.0³,⁸.0¹⁵,²⁰]henicosa-1(14),3,5,7,15,17,19-heptaene
Molecular Weight (g/mol):
290.41
Molecular Formula:
C20H22N2
InChi Key:
YEWGIGCYIAMFMA-UHFFFAOYSA-N
SMILES:
CN1CCC2=C(CC3=CC=CC=C3CC1)NC1=CC=CC=C21
1
–
2
of
2
results
1
Promotions Available
Tocris Bioscience™ LE 300
Tocris Bioscience™ LE 300
2
LE 300, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.