Learn More
Tocris Bioscience™ LE 300
Description
Potent and selective dopamine D1 receptor antagonist (Ki values are 0.08 - 1.9 nM and 6 - 45 nM for D1 and D2 receptors respectively). Also displays moderate affinity for the 5-HT2A receptor (Ki = 20 nM). Active in vivo.
Specifications
Specifications
| CAS | 274694-98-3 |
| Quantity | 10 mg |
| Molecular Formula | C20H22N2 |
| Formula Weight | 290.41 |
| Purity | >98% |
| Molecular Weight (g/mol) | 290.41 |
| Chemical Name or Material | LE 300 |