Learn More
LE 300, TRC
Specifications
Specifications
| Chemical Name or Material | 6,7,8,9,14,15-Hexahydro-7-methyl-5H-indolo[3,2-f][3]benzazecine |
| CAS | 274694-98-3 |
| Molecular Formula | C20H22N2 |
| Quantity | 10 mg |
| Synonym | LE 300, 6,7,8,9,14,15-Hexahydro-7-methyl-5H-indolo[3,2-f][3]benzazecine |
| InChI Formula | InChI=1S/C20H22N2/c1-22-12-10-15-6-2-3-7-16(15)14-20-18(11-13-22)17-8-4-5-9-19(17)21-20/h2-9,21H,10-14H2,1H3 |
| SMILES | CN1CCc2ccccc2Cc3[nH]c4ccccc4c3CC1 |
| Molecular Weight (g/mol) | 290.4 |
| Formula Weight | 290.1783 |