CAS RN 2741-75-5
CAS RN 2741-75-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
(17Beta)-17-Hydroxy-17-methylestr-5(10)-en-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(17Beta)-17-Hydroxy-17-methylestr-5(10)-en-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
(17b)-17-Hydroxy-17-methylestr-5(10)-en-3-one, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.