Learn More
(17Beta)-17-Hydroxy-17-methylestr-5(10)-en-3-one, TRC
Product Code. 30401056
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401056 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30401056 Supplier Toronto Research Chemicals Supplier No. TRCH95070010MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (17beta)-17-Hydroxy-17-methylestr-5(10)-en-3-one
CAS 2741-75-5
Molecular Formula C19H28O2
Quantity 10 mg
Synonym 17beta-Hydroxy-17-methylestr-5(10)-en-3-one, SC 6582
InChI Formula InChI=1S/C19H28O2/c1-18-9-7-15-14-6-4-13(20)11-12(14)3-5-16(15)17(18)8-10-19(18,2)21/h15-17,21H,3-11H2,1-2H3/t15-,16-,17+,18+,19+/m1/s1
SMILES CC1(O)CC[C@@H]2[C@@H]3CCC4=C(CCC(=O)C4)[C@H]3CC[C@]12C
IUPAC Name (8R,9S,13S,14S,17S)-17-hydroxy-13,17-dimethyl-1,2,4,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 288.42
Formula Weight 288.2089