Learn More
(17Beta)-17-Hydroxy-17-methylestr-5(10)-en-3-one, TRC
Specifications
Specifications
| Chemical Name or Material | (17beta)-17-Hydroxy-17-methylestr-5(10)-en-3-one |
| CAS | 2741-75-5 |
| Molecular Formula | C19H28O2 |
| Quantity | 2.5 mg |
| Synonym | 17beta-Hydroxy-17-methylestr-5(10)-en-3-one, SC 6582 |
| InChI Formula | InChI=1S/C19H28O2/c1-18-9-7-15-14-6-4-13(20)11-12(14)3-5-16(15)17(18)8-10-19(18,2)21/h15-17,21H,3-11H2,1-2H3/t15-,16-,17+,18+,19+/m1/s1 |
| SMILES | CC1(O)CC[C@@H]2[C@@H]3CCC4=C(CCC(=O)C4)[C@H]3CC[C@]12C |
| IUPAC Name | (8R,9S,13S,14S,17S)-17-hydroxy-13,17-dimethyl-1,2,4,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 288.42 |
| Formula Weight | 288.2089 |