CAS RN 2724726-85-4
CAS RN 2724726-85-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(3-Chloro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.