Learn More
3-(3-Chloro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Mikromol™
Product Code. 30050376
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050376 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30050376 Supplier Mikromol Supplier No. MM0545.030025
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-(3-Chloro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-(3-Chloro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride
CAS 2724726-85-4
Analyte or Component Names 3-(3-Chloro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H21 Cl N2 . Cl H
Certifications/Compliance ISO 9001
Synonym Clomipramine Hydrochloride Imp. C (EP), Clomipramine Imp. C (EP), Clomipramine USP Related Compound C, Clomipramine USP RC C, 3-(3-Chloro-5H-dibenzo[b,f]azepin-5-yl)-N,N-dimethylpropan-1-amine Hydrochloride, Clomipramine Hydrochloride Imp. C (EP) as Hydrochloride, Clomipramine Related Compound C as Hydrochloride, Clomipramine Hydrochloride Impurity C as Hydrochloride, Clomipramine Impurity C as Hydrochloride
InChI Formula InChI=1S/C19H21ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-11,14H,5,12-13H2,1-2H3;1H
SMILES Cl.CN(C)CCCN1c2ccccc2C=Cc3ccc(Cl)cc13
IUPAC Name 3-(2-chlorobenzo[b][1]benzazepin-11-yl)-N,N-dimethylpropan-1-amine;hydrochloride
Molecular Weight (g/mol) 349.3
Quantity 25 mg
Formula Weight 348.116 g/mol
Show More Show Less
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.