accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3POOH3COH3C

CAS RN 2404-75-3

IUPAC Name: diethyl butylphosphonate
Synonyms: diethyl butylphosphonate
Molecular Weight (g/mol): 194.21
Molecular Formula: C8H19O3P
InChi Key: KEOJOAVYGLGAIM-UHFFFAOYSA-N
SMILES: CCCCP(=O)(OCC)OCC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results