CAS RN 219921-94-5
CAS RN 219921-94-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(S,S’)-3-Methyl-1-(2-piperidinophenyl)butylamine N-Acetyl-glutamate Salt, TRC
CAS: 219921-94-5 Molecular Weight (g/mol): 435.56 IUPAC Name: (2S)-2-acetamidopentanedioic acid;(1S)-3-methyl-1-(2-piperidin-1-ylphenyl)butan-1-amine SMILES: CC(C)C[C@H](N)c1ccccc1N2CCCCC2.CC(=O)N[C@@H](CCC(=O)O)C(=O)O
2
(1S)-3-Methyl-1-[2-(piperidin-1-yl)phenyl]butan-1-amine N-Acetyl-L-glutamate, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.