Learn More
(S,S’)-3-Methyl-1-(2-piperidinophenyl)butylamine N-Acetyl-glutamate Salt, TRC
Product Code. 30133545
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133545 250 mg 250mg
30007718 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133545 Supplier Toronto Research Chemicals Supplier No. TRCM325925250MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 219921-94-5
Molecular Weight (g/mol) 435.56
IUPAC Name (2S)-2-acetamidopentanedioic acid;(1S)-3-methyl-1-(2-piperidin-1-ylphenyl)butan-1-amine
SMILES CC(C)C[C@H](N)c1ccccc1N2CCCCC2.CC(=O)N[C@@H](CCC(=O)O)C(=O)O
Specifications
Molecular Formula C16 H26 N2 . C7 H11 N O5
Synonym (1S)-3-Methyl-1-[2-(piperidin-1-yl)phenyl]butan-1-amine N-Acetyl-L-glutamate, Repaglinide Imp. C (EP) as N-Acetyl-L-glutamate, L-Glutamic acid, N-acetyl-, compd. with (αS)-α-(2-methylpropyl)-2-(1-piperidinyl)benzenemethanamine (1:1), Benzenemethanamine, α-(2-methylpropyl)-2-(1-piperidinyl)-, (αS)-, N-acetyl-L-glutamate (1:1) (9CI)
InChI Formula InChI=1S/C16H26N2.C7H11NO5/c1-13(2)12-15(17)14-8-4-5-9-16(14)18-10-6-3-7-11-18;1-4(9)8-5(7(12)13)2-3-6(10)11/h4-5,8-9,13,15H,3,6-7,10-12,17H2,1-2H3;5H,2-3H2,1H3,(H,8,9)(H,10,11)(H,12,13)/t15-;5-/m00/s1
Quantity 250 mg
Formula Weight 435.2733
Shipping Condition Room Temperature
Chemical Name or Material (1S)-3-Methyl-1-[2-(piperidin-1-yl)phenyl]butan-1-amine N-Acetyl-L-glutamate