CAS RN 21472-86-6
CAS RN 21472-86-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1,2,3-Trichloro-5-methylbenzene, TRC
CAS: 21472-86-6 Molecular Formula: C7 H5 Cl3 Molecular Weight (g/mol): 195.474 Synonym: 1,2,3-Trichloro-5-methylbenzene (ACI),Toluene, 3,4,5-trichloro- (8CI),3,4,5-Trichlorotoluene,3,4,5-Trichlorotoluene IUPAC Name: 1,2,3-trichloro-5-methylbenzene SMILES: Cc1cc(Cl)c(Cl)c(Cl)c1
2
3,4,5-Trichlorotoluene, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality