accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 211555-04-3

CH3ONNHNOHBrOCH3

CAS RN 211555-04-3

IUPAC Name: 2-bromo-4-[(6,7-dimethoxyquinazolin-4-yl)amino]phenol
Synonyms: 2-bromo-4-[(6,7-dimethoxyquinazolin-4-yl)amino]phenol
Molecular Weight (g/mol): 376.21
Molecular Formula: C16H14BrN3O3
InChi Key: CBIAKDAYHRWZCU-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(NC3=CC(Br)=C(O)C=C3)=NC=NC2=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results