Learn More
STEMCELL Technologies WHI-P154
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
- WHI-P154 is an inhibitor of Janus kinase 3 (JAK3) with IC50 values of 28 and 128 μM against human and mouse proteins, respectively (Sudbeck et al.) It has also been reported to show significant inhibition of other kinases, including epidermal growth factor receptor (EGFR) in the nanomolar range (Changelian et al.; Uckun et al.) No significant inhibition of JAK1 or JAK2 has been observed (Sudbeck et al.)
Specifications
Specifications
| CAS | 211555-04-3 |
| Synonym | JAK3 Inhibitor II; Janus-Associated Kinase 3 Inhibitor II |
| Quantity | 10 mg |
| Molecular Formula | C16H14BrN3O3 |
| Purity | ≥98% |