Learn More
WHI-P154, MedChemExpress
Product Code. 16455898
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16455898
Brand: Medchem Express HY13895
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress WHI-P154 is a potent EGFR inhibitor, and also modestly blocks JAK3, with IC50s of 4 nM and 1.8 μM, respectively.
WHI-P154 is a potent EGFR inhibitor, and also modestly blocks JAK3, with IC50s of 4 nM and 1.8 μM, respectively.
Specifications
Chemical Name or Material WHI-P154
CAS 211555-04-3
Color Pink
Purity Grade Notes Research
Molecular Formula C16H14BrN3O3
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (132.91 mM; Need ultrasonic)
SMILES OC1=CC=C(NC2=C3C=C(OC)C(OC)=CC3=NC=N2)C=C1Br
Molecular Weight (g/mol) 376.2
Formula Weight 376.2
Percent Purity 98.81%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease