CAS RN 207853-66-5
CAS RN 207853-66-5
IUPAC Name:
2,5-dibromo-3,6-difluorobenzene-1-sulfonyl chloride
Synonyms:
2,5-dibromo-3,6-difluorobenzenesulfonyl chloride
Molecular Weight (g/mol):
370.39
Molecular Formula:
C6HBr2ClF2O2S
InChi Key:
OTBDNAGWXGDPNX-UHFFFAOYSA-N
SMILES:
FC1=CC(Br)=C(F)C(=C1Br)S(Cl)(=O)=O
1
–
2
of
2
results
1
2,5-Dibromo-3,6-difluorobenzenesulfonyl chloride, 97%, Thermo Scientific™
CAS: 207853-66-5 Molecular Formula: C6HBr2ClF2O2S Molecular Weight (g/mol): 370.39 MDL Number: MFCD00052364 InChI Key: OTBDNAGWXGDPNX-UHFFFAOYSA-N Synonym: 2,5-dibromo-3,6-difluorobenzene-1-sulfonyl chloride,acmc-20aobg,3,6-dibromo-2,5-difluorophenyl chlorosulfone,2,5-dibromo-3,6-difluorophenylsulfonyl chloride PubChem CID: 574297 IUPAC Name: 2,5-dibromo-3,6-difluorobenzenesulfonyl chloride SMILES: FC1=CC(Br)=C(F)C(=C1Br)S(Cl)(=O)=O