missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromo-3,6-difluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11340057 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11340057 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11340057 Supplier Thermo Scientific Acros Supplier No. 357445000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 207853-66-5
CAS Min % 96.0
CAS Max % 100.0
Melting Point 68°C to 73°C
Color Cream
Infrared Spectrum Authentic
Assay Percent Range 96% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Molecular Formula C6HBr2ClF2O2S
MDL Number MFCD00052364
Quantity 500 mg
Synonym 2,5-dibromo-3,6-difluorobenzene-1-sulfonyl chloride, acmc-20aobg, 3,6-dibromo-2,5-difluorophenyl chlorosulfone, 2,5-dibromo-3,6-difluorophenylsulfonyl chloride
InChI Key OTBDNAGWXGDPNX-UHFFFAOYSA-N
SMILES FC1=CC(Br)=C(F)C(=C1Br)S(Cl)(=O)=O
IUPAC Name 2,5-dibromo-3,6-difluorobenzene-1-sulfonyl chloride
Molecular Weight (g/mol) 370.39
PubChem CID 574297
Formula Weight 370.4
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 5-Dibromo-3, 6-difluorobenzenesulfonyl chloride, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.