accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 19179-36-3

IUPAC Name: 3,5-dihydroxybenzonitrile
Synonyms: 3,5-dihydroxybenzonitrile
Molecular Weight (g/mol): 135.12
Molecular Formula: C7H5NO2
InChi Key: ABHOEQJNEOMTEK-UHFFFAOYSA-N
SMILES: OC1=CC(=CC(O)=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results