accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CCH3OCH3CH3

CAS RN 18641-71-9

IUPAC Name: 2,4-dimethylheptan-3-one
Synonyms: 2,4-dimethylheptan-3-one
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: MMEXTUAHJZOQKN-UHFFFAOYNA-N
SMILES: CCCC(C)C(=O)C(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)