Learn More
2,4-Dimethyl-3-heptanone, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dimethyl-3-heptanone |
| CAS | 18641-71-9 |
| Molecular Formula | C9 H18 O |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C9H18O/c1-5-6-8(4)9(10)7(2)3/h7-8H,5-6H2,1-4H3 |
| SMILES | CCCC(C)C(=O)C(C)C |
| IUPAC Name | 2,4-dimethylheptan-3-one |
| Molecular Weight (g/mol) | 142.239 |
| Formula Weight | 142.136 |