Learn More
2,4-Dimethyl-3-heptanone, TRC
Product Code. 30369660
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369660 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30369660 Supplier Toronto Research Chemicals Supplier No. TRCD461868100MG
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4-Dimethyl-3-heptanone
CAS 18641-71-9
Molecular Formula C9 H18 O
Quantity 100 mg
InChI Formula InChI=1S/C9H18O/c1-5-6-8(4)9(10)7(2)3/h7-8H,5-6H2,1-4H3
SMILES CCCC(C)C(=O)C(C)C
IUPAC Name 2,4-dimethylheptan-3-one
Molecular Weight (g/mol) 142.239
Formula Weight 142.136