CAS RN 1805955-65-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-5-fluoro-3-(trifluoromethyl)benzoic Acid, TRC
CAS: 1805955-65-0 Molecular Formula: C8H3O2F4Cl Molecular Weight (g/mol): 242.55 IUPAC Name: 2-chloro-5-fluoro-3-(trifluoromethyl)benzoic acid SMILES: OC(=O)c1cc(F)cc(c1Cl)C(F)(F)F