Learn More
2-Chloro-5-fluoro-3-(trifluoromethyl)benzoic Acid, TRC
Artikelnummer. 30368434
Ansicht ändern
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Packungsgröße:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30368434 10 mg 10mg
30359325 100 mg 100mg
30356835 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30368434 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCC38625310MG
 Restricted Item
Estimated Shipment: 27-08-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 1805955-65-0
Molecular Formula C8H3O2F4Cl
Molecular Weight (g/mol) 242.55
IUPAC Name 2-chloro-5-fluoro-3-(trifluoromethyl)benzoic acid
SMILES OC(=O)c1cc(F)cc(c1Cl)C(F)(F)F
Spezifikation
Quantity 10 mg
InChI Formula InChI=1S/C8H3ClF4O2/c9-6-4(7(14)15)1-3(10)2-5(6)8(11,12)13/h1-2H,(H,14,15)
Formula Weight 241.9758
Chemical Name or Material 2-Chloro-5-fluoro-3-(trifluoromethyl)benzoic Acid