CAS RN 176317-02-5
IUPAC Name:
1-bromo-2,3,4-trifluorobenzene
Synonyms:
1-bromo-2,3,4-trifluorobenzene
Molecular Weight (g/mol):
210.98
Molecular Formula:
C6H2BrF3
InChi Key:
MUUAQFJJUGVBGB-UHFFFAOYSA-N
SMILES:
FC1=CC=C(Br)C(F)=C1F
1
–
2
of
2
results
2
1-Bromo-2,3,4-trifluorobenzene, 99%, Thermo Scientific™
CAS: 176317-02-5 Molecular Formula: C6H2BrF3 Molecular Weight (g/mol): 210.981 MDL Number: MFCD00013248 InChI Key: MUUAQFJJUGVBGB-UHFFFAOYSA-N Synonym: 2,3,4-trifluorobromobenzene,2,3,4-trifluoro bromobenzene,benzene, 1-bromo-2,3,4-trifluoro,1-bromo-2,3,4-trifluoro-benzene,trifluorobromobenzene,pubchem7089,trifluorophenyl bromide,fr bf cf de,acmc-209eb9,ksc497i1f PubChem CID: 2733255 IUPAC Name: 1-bromo-2,3,4-trifluorobenzene SMILES: C1=CC(=C(C(=C1F)F)F)Br