missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3,4-trifluorobenzene, 99%, ACROS Organics™

Product Code. 10083620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10083620 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10083620 Supplier Thermo Scientific Acros Supplier No. 308310010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 176317-02-5
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H2BrF3
Linear Formula BrC6H2F3
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
MDL Number MFCD00013248
Quantity 1 g
Synonym 2,3,4-trifluorobromobenzene, 2,3,4-trifluoro bromobenzene, benzene, 1-bromo-2,3,4-trifluoro, 1-bromo-2,3,4-trifluoro-benzene, trifluorobromobenzene, pubchem7089, trifluorophenyl bromide, fr bf cf de, acmc-209eb9, ksc497i1f
InChI Key MUUAQFJJUGVBGB-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1F)F)F)Br
IUPAC Name 1-bromo-2,3,4-trifluorobenzene
Molecular Weight (g/mol) 210.981
PubChem CID 2733255
Formula Weight 210.98
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 1-Bromo-2, 3, 4-trifluorobenzene, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.