missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2,3,4-trifluorobenzene, 99%, Thermo Scientific™

Product Code. 11458427
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458427 5 g 5g
11448427 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11458427 Supplier Thermo Scientific Alfa Aesar Supplier No. A18514.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Bromo-2,3,4-trifluorobenzene is used in synthesis of 5-nitro-6-methylquinoline carboxylic acid.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 176317-02-5
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.981
MDL Number MFCD00013248
InChI Key MUUAQFJJUGVBGB-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobromobenzene, 2,3,4-trifluoro bromobenzene, benzene, 1-bromo-2,3,4-trifluoro, 1-bromo-2,3,4-trifluoro-benzene, trifluorobromobenzene, pubchem7089, trifluorophenyl bromide, fr bf cf de, acmc-209eb9, ksc497i1f
PubChem CID 2733255
IUPAC Name 1-bromo-2,3,4-trifluorobenzene
SMILES C1=CC(=C(C(=C1F)F)F)Br

Specifications

Density 1.777
Boiling Point 47°C to 49°C (60 mmHg)
Flash Point 62°C (143°F)
Refractive Index 1.486
Quantity 5 g
Beilstein 7805451
Solubility Information Slightly soluble in water.
Formula Weight 210.98
Percent Purity 99%
Chemical Name or Material 1-Bromo-2,3,4-trifluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.