accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 17282-00-7

CH3NHNH2Br

CAS RN 17282-00-7

IUPAC Name: 2-amino-3-bromo-5-methylpyridin-1-ium
Synonyms: 2-amino-3-bromo-5-methylpyridin-1-ium
Molecular Weight (g/mol): 188.05
Molecular Formula: C6H8BrN2
InChi Key: NDPKXEWDWTZBDG-UHFFFAOYSA-O
SMILES: CC1=C[NH+]=C(N)C(Br)=C1
Quantity 
  • (3)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)