Learn More
2-Amino-3-bromo-5-methylpyridine, TRC
Product Code. 30308921
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30308921 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30308921 Supplier Toronto Research Chemicals Supplier No. TRCA6018101G
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Amino-3-bromo-5-methylpyridine
CAS 17282-00-7
Molecular Formula C6H7BrN2
Quantity 1 g
Synonym 3-Bromo-5-methyl-2-pyridinamine, 6-Amino-5-bromo-3-picoline
InChI Formula InChI=1S/C6H7BrN2/c1-4-2-5(7)6(8)9-3-4/h2-3H,1H3,(H2,8,9)
SMILES Cc1cnc(N)c(Br)c1
IUPAC Name 3-bromo-5-methylpyridin-2-amine
Molecular Weight (g/mol) 187.04
Formula Weight 185.9793