accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FHOFCl

CAS RN 164790-68-5

IUPAC Name: 4-chloro-2,6-difluorophenol
Synonyms: 4-chloro-2,6-difluorophenol
Molecular Weight (g/mol): 164.54
Molecular Formula: C6H3ClF2O
InChi Key: BVXYFECYLILSJV-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(Cl)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results