accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FBrFF

CAS RN 157911-55-2

IUPAC Name: 1-(bromomethyl)-2,3,4-trifluorobenzene
Synonyms: 1-(bromomethyl)-2,3,4-trifluorobenzene
Molecular Weight (g/mol): 225.01
Molecular Formula: C7H4BrF3
InChi Key: DGSXDQVPGXFOAN-UHFFFAOYSA-N
SMILES: FC1=CC=C(CBr)C(F)=C1F
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results