missing translation for 'onlineSavingsMsg'
Learn More

2,3,4-Trifluorobenzyl bromide, 97%, Thermo Scientific™

Product Code. 11994541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11994541 5 g 5g
11984541 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11994541 Supplier Thermo Scientific Acros Supplier No. 442260050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 157911-55-2
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.01
InChI Key DGSXDQVPGXFOAN-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzyl bromide, 1-bromomethyl-2,3,4-trifluorobenzene, 2,3,4-trifluorobenzylbromide, 6-bromomethyl-1,2,3-trifluorobenzene, benzene, 1-bromomethyl-2,3,4-trifluoro, alpha-bromo-2,3,4-trifluorotoluene, 2,3,4-trifluoro benzyl bromide, 2,3,4-trifluorophenyl methyl bromide, 4-bromomethyl-1,2,3-trifluorobenzene, 2, 3, 4-trifluorobenzyl bromide
PubChem CID 2777050
IUPAC Name 1-(bromomethyl)-2,3,4-trifluorobenzene
SMILES C1=CC(=C(C(=C1CBr)F)F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 96.5% min. (GC)
Quantity 5 g
Formula Weight 225.01
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3, 4-Trifluorobenzyl bromide
Show More Show Less
compliance-icons
Product Identifier
  • 2,3,4-Trifluorobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.