CAS RN 15421-84-8
CAS RN 15421-84-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Trapidil, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Trapidil, MedChemExpress
MedChemExpress Trapidil is a vasodilator, is an antiplatelet drug with specific platelet-derived growth factor.