Learn More
Trapidil, MedChemExpress
Product Code. 16448648
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16448648
Brand: Medchem Express HYB1016
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Trapidil is a vasodilator, is an antiplatelet drug with specific platelet-derived growth factor.
Trapidil is a vasodilator, is an antiplatelet drug with specific platelet-derived growth factor.
Specifications
Chemical Name or Material Trapidil
Physical Form Solid
CAS 15421-84-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H15N5
For Use With (Application) COVID-19-immunoregulation
Synonym AR-12008
Solubility Information H2O : ≥ 105 mg/mL (511.55 mM)
SMILES CC1=NC2=NC=NN2C(N(CC)CC)=C1
Molecular Weight (g/mol) 205.26
Formula Weight 205.26
Percent Purity 98.0%
Grade Research