CAS RN 152239-46-8
CAS RN 152239-46-8
IUPAC Name:
3-(3-methyl-1,2-thiazol-5-yl)-1-(1-methyl-1H-indol-5-yl)urea
Synonyms:
3-(3-methyl-1,2-thiazol-5-yl)-1-(1-methylindol-5-yl)urea
Molecular Weight (g/mol):
286.35
Molecular Formula:
C14H14N4OS
InChi Key:
USFUFHFQWXDVMH-UHFFFAOYSA-N
SMILES:
CN1C=CC2=CC(NC(=O)NC3=CC(C)=NS3)=CC=C12
1
–
2
of
2
results
1
Tocris Bioscience™ SB 204741
CAS: 152239-46-8 Molecular Formula: C14H14N4OS
2
SB 204741, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.