Learn More
Tocris Bioscience™ SB 204741
GREENER_CHOICE
Product Code. 30001241
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 30001241
Brand: Tocris Bioscience™ 1372/10
Please to purchase this item. Need a web account? Register with us today!
Potent, selective 5-HT2B antagonist
Potent and selective 5-HT2B receptor antagonist (pA2 = 7.95). Displays ≥ 135-fold selectivity over 5-HT2C (pKi = 5.82), 5-HT2A (pKi < 5.2), 5-HT1A, 1D, 1E, 5-HT3 and 5-HT4 receptors.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 152239-46-8
Molecular Formula C14H14N4OS
Specifications
Quantity 10 mg
Formula Weight 286.35
Purity >99%
Molecular Weight (g/mol) 286.353
Chemical Name or Material SB 204741