Learn More
SB 204741, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | N-(1-Methyl-1H-indol-5-yl)-N'-(3-methyl-5-isothiazolyl)urea |
| CAS | 152239-46-8 |
| Molecular Formula | C14H14N4OS |
| Quantity | 500 mg |
| Synonym | SB 204741 |
| InChI Formula | InChI=1S/C14H14N4OS/c1-9-7-13(20-17-9)16-14(19)15-11-3-4-12-10(8-11)5-6-18(12)2/h3-8H,1-2H3,(H2,15,16,19) |
| SMILES | Cc1cc(NC(=O)Nc2ccc3c(ccn3C)c2)sn1 |
| IUPAC Name | 1-(1-methylindol-5-yl)-3-(3-methyl-1,2-thiazol-5-yl)urea |
| Molecular Weight (g/mol) | 286.35 |
| Formula Weight | 286.0888 |