accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 152-47-6

H3CONNHNSOONH2

CAS RN 152-47-6

IUPAC Name: 4-amino-N-(3-methoxypyrazin-2-yl)benzene-1-sulfonamide
Synonyms: longum sulfalene sulfamethoxypyrazine SMP kelfizin kelfizina kelfizine sulfametopyrazine solfametopirazina farmitalia 204/122
Molecular Weight (g/mol): 280.3
Molecular Formula: C11H12N4O3S
InChi Key: KXRZBTAEDBELFD-UHFFFAOYSA-N
SMILES: COC1=NC=CN=C1NS(=O)(=O)C1=CC=C(N)C=C1
Certifications/Compliance 
  • (2)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results