Learn More
Sulfalene, MedChemExpress
Product Code. 16498298
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498298 10 mg 10mg
16474187 50 mg 50mg
16458036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16498298 Supplier Medchem Express Supplier No. HYA0130
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
MedChemExpress Sulfalene (Sulfametopyrazine) is an antimalarial agent. Sulfalene is also a long-acting sulfonamide antibacterial.
Sulfalene (Sulfametopyrazine) is an antimalarial agent. Sulfalene is also a long-acting sulfonamide antibacterial.
Specifica
Chemical Name or Material Sulfalene
Physical Form Solid
CAS 152-47-6
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C11H12N4O3S
For Use With (Application) COVID-19-immunoregulation
Synonym Sulfametopyrazine AS-18908
Solubility Information DMSO : 100 mg/mL (356.76 mM; Need ultrasonic and warming)
SMILES O=S(C1=CC=C(N)C=C1)(NC2=NC=CN=C2OC)=O
Molecular Weight (g/mol) 280.3
Formula Weight 280.3
Percent Purity 99.9%
Grade Research
Vedi altri risultati Mostra meno risultati
Research purposes only
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.