CAS RN 139756-21-1
CAS RN 139756-21-1
IUPAC Name:
5-(2-ethoxyphenyl)-1-methyl-3-propyl-1H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one
Synonyms:
5-(2-ethoxyphenyl)-1-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one
Molecular Weight (g/mol):
312.37
Molecular Formula:
C17H20N4O2
InChi Key:
MXQUEDUMKWBYHI-UHFFFAOYSA-N
SMILES:
CCCC1=NN(C)C2=C1NC(=NC2=O)C1=CC=CC=C1OCC
1
–
1
of
1
results
1
5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.