Learn More
5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one, Mikromol™
Product Code. 30459069
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. 30459069
Brand: Mikromol MM0254.2900254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
CAS 139756-21-1
Analyte or Component Names 5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
Shipping Condition Room Temperature
Molecular Formula C17 H20 N4 O2
Certifications/Compliance ISO 17025
Synonym 5-(2-Ethoxyphenyl)-1-methyl-3n-propyl-1,6-dihydro-7H-pyrazolo[4,3-d]pyrimidin-7-one
InChI Formula InChI=1S/C17H20N4O2/c1-4-8-12-14-15(21(3)20-12)17(22)19-16(18-14)11-9-6-7-10-13(11)23-5-2/h6-7,9-10H,4-5,8H2,1-3H3,(H,18,19,22)
Solution Type Neat
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3ccccc3OCC
IUPAC Name 5-(2-ethoxyphenyl)-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
Molecular Weight (g/mol) 312.37
Quantity 4 x 25 mg
Formula Weight 312.1586 g/mol