accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 13925-00-3

H3CNN

CAS RN 13925-00-3

IUPAC Name: 2-ethylpyrazine
Synonyms: 2-ethyl pyrazine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: KVFIJIWMDBAGDP-UHFFFAOYSA-N
SMILES: CCC1=CN=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results