Learn More
2-Ethylpyrazine, 99%, Thermo Scientific™
Product Code. 11435093
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11435093
Brand: Thermo Scientific Alfa Aesar A12759.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13925-00-3
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006149
InChI Key KVFIJIWMDBAGDP-UHFFFAOYSA-N
Synonym ethylpyrazine, pyrazine, ethyl, 2-ethyl-1,4-diazine, 2-ethyl pyrazine, moldin, pyrazine, 2-ethyl, fema no. 3281, unii-0qo4luv16z, 2-ethyl-pyrazine, ccris 2931
PubChem CID 26331
ChEBI CHEBI:73232
IUPAC Name 2-ethylpyrazine
SMILES CCC1=CN=CC=N1
Specifications
Density 0.988
Boiling Point 152°C to 153°C
Flash Point 42°C (107°F)
Refractive Index 1.498
Quantity 25 g
UN Number UN1993
Beilstein 108200
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2-Ethylpyrazine