Learn More
Ethylpyrazine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2-Ethylpyrazine |
| CAS | 13925-00-3 |
| Molecular Formula | C6 H8 N2 |
| Quantity | 1 g |
| Synonym | Pyrazine, 2-ethyl-, Pyrazine, ethyl- (6CI,7CI,8CI,9CI), 2-Ethylpyrazine, Ethylpyrazine, C-Ethylpyrazine |
| InChI Formula | InChI=1S/C6H8N2/c1-2-6-5-7-3-4-8-6/h3-5H,2H2,1H3 |
| SMILES | CCc1cnccn1 |
| IUPAC Name | 2-ethylpyrazine |
| Molecular Weight (g/mol) | 108.14 |
| Formula Weight | 108.0687 |