CAS RN 1380424-42-9
CAS RN 1380424-42-9
IUPAC Name:
1,1,1,3,3,3-hexafluoropropan-2-yl 4-[bis(2H-1,3-benzodioxol-5-yl)(hydroxy)methyl]piperidine-1-carboxylate
Synonyms:
1,1,1,3,3,3-hexafluoropropan-2-yl 4-[bis(2H-1,3-benzodioxol-5-yl)(hydroxy)methyl]piperidine-1-carboxylate
Molecular Weight (g/mol):
549.42
Molecular Formula:
C24H21F6NO7
InChi Key:
SXHQLPHDBLTFPM-UHFFFAOYSA-N
SMILES:
OC(C1CCN(CC1)C(=O)OC(C(F)(F)F)C(F)(F)F)(C1=CC2=C(OCO2)C=C1)C1=CC2=C(OCO2)C=C1
1
–
4
of
4
results
1
KML-29, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
KML-29, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
4
KML29, MedChemExpress
MedChemExpress KML29 is an extremely selective, orally active and irreversible MAGL inhibitor, with IC50 values of 15 nM, 43 nM and 5.9 nM for mouse, rat and human MAGL, respectively. KML29 exhibits minimal cross-reactivity toward other central and peripheral serine hydrolases, including no detectable activity against FAAH.