missing translation for 'onlineSavingsMsg'
Learn More

KML29, MedChemExpress

Product Code. p-200097979
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16487668

Brand: Medchem Express HY18977

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KML29 is an extremely selective, orally active and irreversible MAGL inhibitor, with IC50 values of 15 nM, 43 nM and 5.9 nM for mouse, rat and human MAGL, respectively. KML29 exhibits minimal cross-reactivity toward other central and peripheral serine hydrolases, including no detectable activity against FAAH.

KML29 is an extremely selective, orally active and irreversible MAGL inhibitor, with IC50 values of 15 nM, 43 nM and 5.9 nM for mouse, rat and human MAGL, respectively. KML29 exhibits minimal cross-reactivity toward other central and peripheral serine hydrolases, including no detectable activity against FAAH.

Specifications

Chemical Name or Material KML29
CAS 1380424-42-9
Color White
Purity Grade Notes Research
Molecular Formula C24H21F6NO7
Quantity 10 mg
Solubility Information DMSO : 50 mg/mL (91.01 mM; Need ultrasonic)
SMILES O=C(N1CCC(C(C2=CC=C(OCO3)C3=C2)(C4=CC=C(OCO5)C5=C4)O)CC1)OC(C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 549.42
Formula Weight 549.42
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.