Learn More
KML-29, TRC
Product Code. 30404938
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404938 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30404938 Supplier Toronto Research Chemicals Supplier No. TRCK64850025MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material KML29
CAS 1380424-42-9
Molecular Formula C24 H21 F6 N O7
Quantity 25 mg
Synonym 4-[Bis(1,3-benzodioxol-5-yl)hydroxymethyl]-1-piperidinecarboxylic Acid 2,2,2-Trifluoro-1-(trifluoromethyl)ethyl Ester, 1,1,1,3,3,3-Hexafluoropropan-2-yl 4-(Bis(benzo[d][1,3]dioxol-5-yl)(hydroxy)methyl)piperidine-1-carboxylate, KML29
InChI Formula InChI=1S/C24H21F6NO7/c25-23(26,27)20(24(28,29)30)38-21(32)31-7-5-13(6-8-31)22(33,14-1-3-16-18(9-14)36-11-34-16)15-2-4-17-19(10-15)37-12-35-17/h1-4,9-10,13,20,33H,5-8,11-12H2
SMILES OC(C1CCN(CC1)C(=O)OC(C(F)(F)F)C(F)(F)F)(c2ccc3OCOc3c2)c4ccc5OCOc5c4
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-yl 4-[bis(1,3-benzodioxol-5-yl)-hydroxymethyl]piperidine-1-carboxylate
Molecular Weight (g/mol) 549.42
Formula Weight 549.1222