accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNNONHOHN

CAS RN 137592-45-1

IUPAC Name: 3-(1H-indol-3-yl)-4-{1-[2-(1-methylpyrrolidin-2-yl)ethyl]-1H-indol-3-yl}-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 3-(1H-indol-3-yl)-4-{1-[2-(1-methylpyrrolidin-2-yl)ethyl]indol-3-yl}-1H-pyrrole-2,5-dione
Molecular Weight (g/mol): 438.53
Molecular Formula: C27H26N4O2
InChi Key: LBFDERUQORUFIN-UHFFFAOYNA-N
SMILES: CN1CCCC1CCN1C=C(C2=CC=CC=C12)C1=C(C(=O)NC1=O)C1=CNC2=CC=CC=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results