missing translation for 'onlineSavingsMsg'
Learn More

Bisindolylmaleimide II, Tocris Bioscience™
GREENER_CHOICE

Product Code. 30001473
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001473 50 mg 50mg
30001466 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001473 Supplier Tocris Bioscience Supplier No. 4128/50

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Potent PKC inhibitor and nicotinic receptor antagonist

Potent, ATP-competitive protein kinase C (PKC) inhibitor (IC50 = 0.01 μM). Displays selectivity for PKC over protein kinase A (PKA) and phosphorylase kinase (PK) (IC50 values are 0.75 and 2μM for PK and PKA respectively). Also displays potent, noncompetitive antagonism at nicotinic cholinergic receptors (IC50 ∽ 0.03 μM for inhibition of catecholamine secretion in nicotine-stimulated PC-12 cells).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137592-45-1
Molecular Formula C27H26N4O2
Molecular Weight (g/mol) 438.531
InChI Key LBFDERUQORUFIN-UHFFFAOYSA-N
Synonym bisindolylmaleimide ii, 2-1-2-1-methylpyrrolidino ethyl-1h-indol-3-yl-3-1h-indol-3-yl maleimide, bisindolyl deriv. 22, bim-2, bisindolylmaleimide ii mixture of 2 isomers, 3-1h-indol-3-yl-4-1-2-1-methylpyrrolidin-2-yl ethyl indol-3-yl pyrrole-2,5-dione, 3-1h-indol-3-yl-4-1-2-1-methyl-2-pyrrolidinyl ethyl-1h-indol-3-yl-1h-pyrrole-2,5-dione, 3-1h-indol-3-yl-4-1-2-1-methylpyrrolidin-2-yl ethyl-1h-indol-3-yl-2,5-dihydro-1h-pyrrole-2,5-dione, 3-1h-indol-3-yl-4-1-2-1-methylpyrrolidin-2-yl ethyl indol-3-yl-1h-pyrrole-2,5-dione
PubChem CID 2397
IUPAC Name 3-(1H-indol-3-yl)-4-[1-[2-(1-methylpyrrolidin-2-yl)ethyl]indol-3-yl]pyrrole-2,5-dione
SMILES CN1CCCC1CCN2C=C(C3=CC=CC=C32)C4=C(C(=O)NC4=O)C5=CNC6=CC=CC=C65

Specifications

Quantity 50 mg
Formula Weight 438.52
Percent Purity >97%
Chemical Name or Material Bisindolylmaleimide II
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.