accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CaH3CCH3N(R)OH(R)OHOOFONHH3CCH3N(R)OH(R)OHOOFONH

CAS RN 134523-03-8

IUPAC Name: calcium bis((3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-(propan-2-yl)-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoate)
Synonyms: lipitor liprimar atorvastan
Molecular Weight (g/mol): 1155.36
Molecular Formula: C66H68CaF2N4O10
InChi Key: FQCKMBLVYCEXJB-MNSAWQCASA-L
SMILES: [Ca++].CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1.CC(C)C1=C(C(=O)NC2=CC=CC=C2)C(=C(N1CC[C@@H](O)C[C@@H](O)CC([O-])=O)C1=CC=C(F)C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (3)
  • (5)
  • (5)
  • (5)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (5)
  • (1)
  • (1)
  • (1)