CAS RN 1343038-33-4
CAS RN 1343038-33-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Chloro-2-fluoro-N-methylbenzamide, TRC
CAS: 1343038-33-4 Molecular Formula: C8 H7 Cl F N O Molecular Weight (g/mol): 187.599 Synonym: Apalutamide Impurity 27 IUPAC Name: 4-chloro-2-fluoro-N-methylbenzamide SMILES: CNC(=O)c1ccc(Cl)cc1F